Search
Search

RCN1| Gene Symbol: RCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102360.4 | NP_173920.1 | 12 - 13 | 1816 | 66 | |
| GenBank mRNA | AY120757.1 | - | 12 - 13 | 1758 | 66 | |
| BT000108.1 | - | 11 - 12 | 1641 | 66 | ||
Set Membership: |
|