Search
Search

AT1G28060| Gene Symbol: AT1G28060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332796.1 | NP_001319094.1 | 4 - 5 | 1322 | 129 | |
| NM_001332797.1 | NP_001321377.1 | 3 - 4 | 1286 | 129 | ||
| NM_102571.3 | NP_174127.1 | 3 - 4 | 1302 | 129 | ||
| GenBank mRNA | AY126993.1 | - | 3 - 4 | 1167 | 129 | |
Set Membership: |
|