Search
Search

FIM5| Gene Symbol: FIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122961.4 | NP_198420.1 | 6 - 7 | 940 | 108 | |
| GenBank mRNA | AY045780.1 | - | 6 - 7 | 918 | 108 | |
| AY142570.1 | - | 5 - 6 | 715 | 108 | ||
Set Membership: |
|