Search
Search

RAD23D| Gene Symbol: RAD23D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085211.1 | NP_001078680.1 | 11 - 12 | 1096 | 67 | |
| NM_123208.2 | NP_198663.1 | 11 - 12 | 1112 | 67 | ||
| GenBank mRNA | AB109198.1 | - | 11 - 12 | 1014 | 67 | |
| AY058196.1 | - | 11 - 12 | 1092 | 67 | ||
| AY081835.1 | - | 11 - 12 | 1014 | 67 | ||
| AY087564.1 | - | 11 - 12 | 1092 | 67 | ||
Set Membership: |
|