Search
Search

NRPB12| Gene Symbol: NRPB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123466.3 | NP_198917.1 | 2 - 3 | 267 | 76 | |
| GenBank mRNA | AY045954.1 | - | 2 - 3 | 226 | 76 | |
| AY079307.1 | - | 2 - 3 | 136 | 76 | ||
| AY084743.1 | - | 2 - 3 | 253 | 76 | ||
Set Membership: |
|