Search
Search

ATS| Gene Symbol: ATS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125891.2 | NP_001119363.1 | 2 - 3 | 569 | 106 | |
| NM_123627.3 | NP_199077.1 | 2 - 3 | 617 | 106 | ||
| GenBank mRNA | AY048691.1 | - | 2 - 3 | 468 | 106 | |
| BT026053.1 | - | 2 - 3 | 460 | 106 | ||
Set Membership: |
|