Search
Search

COP8| Gene Symbol: COP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123663.3 | NP_199111.1 | 10 - 11 | 1126 | 86 | |
| GenBank mRNA | AF395060.1 | - | 10 - 11 | 959 | 86 | |
| AK176360.1 | - | 10 - 11 | 1041 | 86 | ||
| BT025548.1 | - | 10 - 11 | 959 | 86 | ||
Set Membership: |
|