Search
Search

UAH| Gene Symbol: UAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123726.4 | NP_199173.2 | 4 - 5 | 712 | 72 | |
| GenBank mRNA | AK230338.1 | - | 1678 | 72 | ||
| AY074343.1 | - | 4 - 5 | 676 | 72 | ||
| AY091387.1 | - | 4 - 5 | 501 | 72 | ||
Set Membership: |
|