Search
Search

AGO10| Gene Symbol: AGO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203535.1 | NP_001190464.1 | 13 - 14 | 2601 | 147 | |
| NM_001344505.1 | NP_001331600.1 | 12 - 13 | 2524 | 147 | ||
| NM_001344506.1 | NP_001331599.1 | 12 - 13 | 2341 | 147 | ||
| NM_123748.3 | NP_199194.1 | 12 - 13 | 2396 | 147 | ||
Set Membership: |
|