Search
Search

HOT5| Gene Symbol: HOT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203539.1 | NP_001190468.1 | 8 - 9 | 1168 | 78 | |
| NM_123761.4 | NP_199207.1 | 8 - 9 | 1098 | 78 | ||
| GenBank mRNA | AK226412.1 | - | 8 - 9 | 1089 | 78 | |
| AY039601.1 | - | 8 - 9 | 1074 | 78 | ||
| AY087250.1 | - | 8 - 9 | 1086 | 78 | ||
| BT010169.1 | - | 8 - 9 | 1023 | 78 | ||
Set Membership: |
|