Search
Search

GLK2| Gene Symbol: GLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344537.1 | NP_001331906.1 | 1 - 2 | 616 | 118 | |
| NM_123786.4 | NP_199232.1 | 1 - 2 | 597 | 118 | ||
| GenBank mRNA | AB062490.1 | - | 1 - 2 | 443 | 118 | |
| AY026773.1 | - | 1 - 2 | 443 | 118 | ||
Set Membership: |
|