Search
Search

KASI| Gene Symbol: KASI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036941.1 | NP_001032018.1 | 5 - 6 | 1125 | 61 | |
| NM_001203550.1 | NP_001190479.1 | 4 - 5 | 1297 | 61 | ||
| NM_123998.3 | NP_199441.1 | 4 - 5 | 1394 | 61 | ||
| GenBank mRNA | AY037261.1 | - | 4 - 5 | 1284 | 61 | |
| AY087843.1 | - | 4 - 5 | 1259 | 61 | ||
| AY094005.1 | - | 4 - 5 | 1184 | 61 | ||
| AY123979.1 | - | 4 - 5 | 1288 | 61 | ||
Set Membership: |
|