Search
Search

GAUT4| Gene Symbol: GAUT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124152.3 | NP_568688.1 | 7 - 8 | 1113 | 86 | |
| GenBank mRNA | AY056202.1 | - | 7 - 8 | 1061 | 86 | |
| AY091394.1 | - | 7 - 8 | 996 | 86 | ||
| KJ138647.1 | - | 7 - 8 | 996 | 86 | ||
Set Membership: |
|