Search
Search

SLD5| Gene Symbol: SLD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125927.2 | NP_001119399.1 | 4 - 5 | 919 | 97 | |
| NM_124278.4 | NP_199712.2 | 5 - 6 | 784 | 97 | ||
| GenBank mRNA | AK221044.1 | - | 5 - 6 | 784 | 97 | |
| BT010406.1 | - | 5 - 6 | 551 | 97 | ||
Set Membership: |
|