Search
Search

AT5G49900| Gene Symbol: AT5G49900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344862.1 | NP_001331234.1 | 9 - 10 | 1911 | 76 | |
| NM_001344863.1 | NP_001331233.1 | 19 - 20 | 2871 | 76 | ||
| NM_124368.4 | NP_199801.2 | 19 - 20 | 2786 | 76 | ||
| GenBank mRNA | BT002482.1 | - | 19 - 20 | 2626 | 76 | |
| BT010551.1 | - | 18 - 19 | 2575 | 76 | ||
Set Membership: |
|