Search
Search

PAP27| Gene Symbol: PAP27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344892.1 | NP_001330206.1 | 1 - 2 | 407 | 86 | |
| NM_124422.4 | NP_199851.2 | 1 - 2 | 459 | 86 | ||
| GenBank mRNA | AY842027.1 | - | 1 - 2 | 169 | 86 | |
Set Membership: |
|