Search
Search

PABN1| Gene Symbol: PABN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203582.1 | NP_001190511.1 | 4 - 5 | 545 | 71 | |
| NM_001203583.1 | NP_001190512.1 | 2 - 3 | 96 | 71 | ||
| NM_124491.3 | NP_568751.1 | 3 - 4 | 550 | 71 | ||
| GenBank mRNA | AY054186.1 | - | 3 - 4 | 391 | 71 | |
| AY057580.1 | - | 3 - 4 | 378 | 71 | ||
| AY079392.1 | - | 3 - 4 | 332 | 71 | ||
| AY116673.1 | - | 3 - 4 | 375 | 71 | ||
Set Membership: |
|