Search
Search

CPSF160| Gene Symbol: CPSF160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344949.1 | NP_001331731.1 | 11 - 12 | 1742 | 99 | |
| NM_001344950.1 | NP_001331729.1 | 11 - 12 | 1718 | 99 | ||
| NM_001344951.1 | NP_001331730.1 | 11 - 12 | 1643 | 99 | ||
| NM_124545.3 | NP_199979.2 | 11 - 12 | 1746 | 99 | ||
Set Membership: |
|