Search
Search

PGM| Gene Symbol: PGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124561.3 | NP_199995.1 | 20 - 21 | 2091 | 95 | |
| GenBank mRNA | AF216580.1 | - | 20 - 21 | 1806 | 95 | |
| AY099708.1 | - | 20 - 21 | 1762 | 95 | ||
| AY128901.1 | - | 20 - 21 | 1697 | 95 | ||
Set Membership: |
|