Search
Search

PGM| Gene Symbol: PGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124561.3 | NP_199995.1 | 2 - 3 | 661 | 99 | |
| GenBank mRNA | AF216580.1 | - | 2 - 3 | 376 | 99 | |
| AY099708.1 | - | 2 - 3 | 332 | 99 | ||
| AY128901.1 | - | 2 - 3 | 267 | 99 | ||
Set Membership: |
|