Search
Search

LTI65| Gene Symbol: LTI65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036984.1 | NP_001032061.1 | 2 - 3 | 384 | 71 | |
| NM_124609.4 | NP_200043.2 | 2 - 3 | 422 | 71 | ||
| GenBank mRNA | AY081282.1 | - | 2 - 3 | 386 | 71 | |
| AY128731.1 | - | 2 - 3 | 238 | 71 | ||
Set Membership: |
|