Search
Search

USP| Gene Symbol: USP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124635.4 | NP_568775.1 | 11 - 12 | 1308 | 138 | |
| GenBank mRNA | AF360236.1 | - | 11 - 12 | 1171 | 138 | |
| AY040035.1 | - | 10 - 11 | 985 | 138 | ||
| DQ324529.1 | - | 10 - 11 | 985 | 138 | ||
Set Membership: |
|