Search
Search

OMT1| Gene Symbol: OMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124796.4 | NP_200227.1 | 1 - 2 | 589 | 82 | |
| GenBank mRNA | AY062837.1 | - | 1 - 2 | 494 | 82 | |
| AY081565.1 | - | 1 - 2 | 416 | 82 | ||
| AY087297.1 | - | 1 - 2 | 494 | 82 | ||
Set Membership: |
|