Search
Search

ERO2| Gene Symbol: ERO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202779.1 | NP_001189708.1 | 2 - 3 | 368 | 73 | |
| NM_129454.4 | NP_181430.2 | 2 - 3 | 481 | 73 | ||
| NM_201908.1 | NP_973637.1 | 2 - 3 | 312 | 73 | ||
| GenBank mRNA | AJ551466.1 | - | 2 - 3 | 312 | 73 | |
| BT022038.1 | - | 2 - 3 | 333 | 73 | ||
| BT023745.1 | - | 2 - 3 | 312 | 73 | ||
Set Membership: |
|