Search
Search

NRPD1B| Gene Symbol: NRPD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336818.1 | NP_001323985.1 | 16 - 17 | 5795 | 60 | |
| NM_129561.4 | NP_181532.2 | 17 - 18 | 5668 | 60 | ||
| GenBank mRNA | AY826516.1 | - | 16 - 17 | 5394 | 60 | |
| AY927744.1 | - | 16 - 17 | 5394 | 60 | ||
Set Membership: |
|