Search
Search

AT2G40110| Gene Symbol: AT2G40110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336822.1 | NP_001324481.1 | 2 - 3 | 473 | 100 | |
| NM_001336823.1 | NP_001324480.1 | 2 - 3 | 431 | 100 | ||
| NM_129569.5 | NP_181540.1 | 2 - 3 | 473 | 100 | ||
| NM_201916.2 | NP_973645.1 | 2 - 3 | 431 | 100 | ||
| GenBank mRNA | AK317386.1 | - | 2 - 3 | 279 | 100 | |
| AY085617.1 | - | 2 - 3 | 337 | 100 | ||
| BT001947.1 | - | 2 - 3 | 145 | 100 | ||
Set Membership: |
|