Search
Search

WRKY55| Gene Symbol: WRKY55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336861.1 | NP_001324304.1 | 2 - 3 | 679 | 106 | |
| NM_129636.3 | NP_181606.1 | 2 - 3 | 580 | 106 | ||
| GenBank mRNA | AB493584.1 | - | 2 - 3 | 580 | 106 | |
| DQ056575.1 | - | 2 - 3 | 580 | 106 | ||
Set Membership: |
|