Search
Search

DPE2| Gene Symbol: DPE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129647.4 | NP_181616.3 | 16 - 17 | 2193 | 148 | |
| GenBank mRNA | AY081315.1 | - | 16 - 17 | 1984 | 148 | |
| BT010364.1 | - | 15 - 16 | 1935 | 148 | ||
Set Membership: |
|