Search
Search

CRK1| Gene Symbol: CRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129679.2 | NP_181647.1 | 8 - 9 | 1757 | 117 | |
| GenBank mRNA | AF153351.1 | - | 8 - 9 | 1465 | 117 | |
| AF435448.1 | - | 8 - 9 | 1465 | 117 | ||
Set Membership: |
|