Search
Search

NUDX23| Gene Symbol: NUDX23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336950.1 | NP_001324265.1 | 5 - 6 | 832 | 243 | |
| NM_129770.4 | NP_565965.1 | 5 - 6 | 1021 | 142 | ||
| GenBank mRNA | AK221588.1 | - | 5 - 6 | 824 | 142 | |
| BT025803.1 | - | 5 - 6 | 142 | |||
Set Membership: |
|