Search
Search

HOL1| Gene Symbol: HOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337065.1 | NP_001318420.1 | 1 - 2 | 181 | 76 | |
| NM_129953.3 | NP_181919.1 | 1 - 2 | 511 | 76 | ||
| GenBank mRNA | AY044314.1 | - | 1 - 2 | 104 | 76 | |
| AY086360.1 | - | 1 - 2 | 178 | 76 | ||
Set Membership: |
|