Search
Search

GL3| Gene Symbol: GL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344395.1 | NP_001332706.1 | 2 - 3 | 772 | 93 | |
| NM_001344396.1 | NP_001332705.1 | 3 - 4 | 628 | 93 | ||
| NM_148067.4 | NP_680372.1 | 2 - 3 | 407 | 93 | ||
| GenBank mRNA | AF246291.1 | - | 2 - 3 | 405 | 93 | |
Set Membership: |
|