Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› At02329915_s1
See other ACT2 GE Assays ›
Gene Symbol:
ACT2
Gene Name:
actin 2
Gene Aliases:
AT3G18780, DEFORMED ROOT HAIRS 1, DER1, ENHANCER OF LRX1 2, ENL2, FIZ2, FRIZZY AND KINKED SHOOTS 2, LIGHT STRESS-REGULATED 2, LSR2, actin 2
Chromosome Location:
Chr.3: 6474710 - 6477204 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID
At02329915_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ACT2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001338358.1
NP_001319585.1
3
1575
124
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Endogenous Controls
Detect Genomic DNA
Arabidopsis thaliana
Inventoried
Both primers and probe map within a single exon
Panther Classification:
Molecular Function -
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
response to high light intensity
response to cytokinin
root epidermal cell differentiation
response to red light
response to far red light
root hair elongation
root hair cell tip growth
structural constituent of cytoskeleton
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline