Search
Search

AT2G17360| Gene Symbol: AT2G17360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202610.1 | NP_001189539.1 | 3 - 4 | 380 | 125 | |
| NM_127291.4 | NP_565414.1 | 3 - 4 | 420 | 125 | ||
| GenBank mRNA | AF370469.1 | - | 3 - 4 | 335 | 125 | |
| AY035131.1 | - | 3 - 4 | 334 | 125 | ||
| AY062983.1 | - | 2 - 3 | 262 | 125 | ||
| AY064625.1 | - | 2 - 3 | 262 | 125 | ||
| AY084230.1 | - | 3 - 4 | 375 | 125 | ||
Set Membership: |
|