Search
Search

H2AXA| Gene Symbol: H2AXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100760.5 | NP_172363.1 | 1 - 2 | 629 | 117 | |
| GenBank mRNA | AF412092.1 | - | 1 - 2 | 307 | 117 | |
| AY093975.1 | - | 1 - 2 | 205 | 117 | ||
Set Membership: |
|