Search
Search

RPP5| Gene Symbol: RPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341165.1 | NP_001328022.1 | 1 - 2 | 645 | 136 | |
| NM_001341166.1 | NP_001328019.1 | 1 - 2 | 611 | 136 | ||
| NM_001341167.1 | NP_001328020.1 | 1 - 2 | 611 | 136 | ||
| NM_001341168.1 | NP_001328021.1 | 1 - 2 | 611 | 136 | ||
| NM_117798.4 | NP_193428.2 | 1 - 2 | 611 | 136 | ||
| NM_179067.2 | NP_849398.1 | 1 - 2 | 899 | 136 | ||
Set Membership: |
|