Search
Search

NIT1| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084765.1 | NP_001078234.1 | 4 - 5 | 916 | 170 | |
| NM_001339127.1 | NP_001319680.1 | 3 - 4 | 1047 | 170 | ||
| NM_001339128.1 | NP_001325848.1 | 3 - 4 | 1009 | 170 | ||
| NM_180680.3 | NP_851011.1 | 4 - 5 | 1156 | 170 | ||
| GenBank mRNA | AB493638.1 | - | 4 - 5 | 879 | 170 | |
| AY042847.1 | - | 4 - 5 | 914 | 170 | ||
| AY084633.1 | - | 3 - 4 | 798 | 170 | ||
| BT000040.1 | - | 4 - 5 | 879 | 170 | ||
| DQ446730.1 | - | 4 - 5 | 879 | 170 | ||
Set Membership: |
|