Search
Search

CYP71B22| Gene Symbol: CYP71B22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113527.2 | NP_189251.1 | 1 - 2 | 986 | 117 | |
| GenBank mRNA | AF360271.1 | - | 1 - 2 | 953 | 117 | |
| AY040080.1 | - | 1 - 2 | 888 | 117 | ||
Set Membership: |
|