Search
Search

GATA19| Gene Symbol: GATA19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119825.3 | NP_195380.1 | 1 - 2 | 622 | 165 | |
| GenBank mRNA | AB493721.1 | - | 1 - 2 | 285 | 165 | |
| BT029506.1 | - | 1 - 2 | 285 | 165 | ||
Set Membership: |
|