Search
Search

BIP1| Gene Symbol: BIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122737.4 | NP_198206.1 | 5 - 6 | 1788 | 166 | |
| GenBank mRNA | BT000453.1 | - | 5 - 6 | 1571 | 166 | |
| BT001212.1 | - | 5 - 6 | 1504 | 166 | ||
Set Membership: |
|