Search
Search

MLP329| Gene Symbol: MLP329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126214.5 | NP_565265.1 | 1 - 2 | 334 | 177 | |
| GenBank mRNA | AB493544.1 | - | 1 - 2 | 188 | 177 | |
| AY052300.1 | - | 1 - 2 | 247 | 177 | ||
| AY061925.1 | - | 1 - 2 | 188 | 177 | ||
Set Membership: |
|