Search
Search

AT1G26270| Gene Symbol: AT1G26270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102391.3 | NP_564242.1 | 2 | 2460 | 159 | |
| GenBank mRNA | AY035014.1 | - | 2 | 2360 | 159 | |
| AY058170.1 | - | 2 | 2352 | 159 | ||
| AY075684.1 | - | 2 | 2352 | 159 | ||
Set Membership: |
|