Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124986.1 | NP_001118458.1 | 2 | 1607 | 156 | |
NM_129243.4 | NP_181225.1 | 2 | 1575 | 156 | ||
GenBank mRNA | AF367310.1 | - | 2 | 1569 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |