Search
Search

RGP5| Gene Symbol: RGP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121657.2 | NP_197155.1 | 2 | 1217 | 139 | |
| NM_180500.3 | NP_850831.1 | 2 | 1303 | 139 | ||
| GenBank mRNA | AY088511.1 | - | 2 | 1179 | 139 | |
| AY091141.1 | - | 2 | 1161 | 139 | ||
Set Membership: |
|