Search
Search

HSFA6B| Gene Symbol: HSFA6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113182.2 | NP_188922.1 | 1 - 2 | 547 | 148 | |
| GenBank mRNA | AY045974.1 | - | 1 - 2 | 516 | 148 | |
| AY079374.1 | - | 1 - 2 | 355 | 148 | ||
Set Membership: |
|