Search
Search

LDL3| Gene Symbol: LDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341093.1 | NP_001319957.1 | 6 - 7 | 4436 | 78 | |
| NM_001341094.1 | NP_001328665.1 | 6 - 7 | 4445 | 78 | ||
| NM_001341095.1 | NP_001328666.1 | 6 - 7 | 4445 | 78 | ||
| NM_001341096.1 | NP_001328664.1 | 6 - 7 | 4388 | 78 | ||
| NM_117726.5 | NP_193364.5 | 5 - 6 | 4032 | 78 | ||
Set Membership: |
|