Search
Search

MYB73| Gene Symbol: MYB73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119889.2 | NP_195443.1 | 1 | 747 | 111 | |
| GenBank mRNA | AY063912.1 | - | 1 | 747 | 111 | |
| AY091267.1 | - | 1 | 635 | 111 | ||
| AY519613.1 | - | 1 | 635 | 111 | ||
Set Membership: |
|