Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ba00005007_po
Gene Symbol:
Bacteria
Gene Name:
Class A extended-spectrum beta-lactamase VEB|PER|GES
Gene Aliases:
Bacteria, blaGES-1, blaGES-10, blaGES-11, blaGES-12, blaGES-13, blaGES-14, blaGES-15, blaGES-16, blaGES-17, blaGES-18, blaGES-19, blaGES-2, blaGES-20, blaGES-21, blaGES-22, blaGES-24, blaGES-25, blaGES-26, blaGES-27, blaGES-28, blaGES-29, blaGES-3, blaGES-30, blaGES-31, blaGES-33, blaGES-34, blaGES-35, blaGES-36, blaGES-37, blaGES-38, blaGES-39, blaGES-4, blaGES-40, blaGES-42, blaGES-43, blaGES-45, blaGES-46, blaGES-47, blaGES-48, blaGES-5, blaGES-6, blaGES-7, blaGES-8, blaGES-9, blaPER-1, blaPER-10, blaPER-11, blaPER-13, blaPER-15, blaPER-16, blaPER-3, blaPER-4, blaPER-5, blaPER-7, blaPER-8, blaPER-9, blaVEB-1, blaVEB-10, blaVEB-11, blaVEB-12, blaVEB-13, blaVEB-15, blaVEB-16, blaVEB-17, blaVEB-18, blaVEB-19, blaVEB-2, blaVEB-20, blaVEB-21, blaVEB-22, blaVEB-23, blaVEB-24, blaVEB-25, blaVEB-26, blaVEB-27, blaVEB-3, blaVEB-4, blaVEB-5, blaVEB-6, blaVEB-7, blaVEB-8, blaVEB-9, carbapenem-hydrolyzing class A beta-lactamase (blaGES), class A beta-lactamase (blaGES), class A extended-spectrum beta-lactamase (blaGES), class A extended-spectrum beta-lactamase (blaPER), class A extended-spectrum beta-lactamase (blaVEB), inhibitor-resistant carbapenem-hydrolyzing class A beta-lactamase (blaGES), inhibitor-resistant class A extended-spectrum beta-lactamase (blaPER), inhibitor-resistant class A extended-spectrum beta-lactamase (blaVEB), blaGES,blaPER,blaVEB
Chromosome Location:
-
Species:
Bacteria
Species Specific ID (Flybase ID):
-
Assay ID
Ba00005007_po
Best Coverage
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Updated Mapping Information
This pooled assay is composed of the following assays: Ba04646140_s1, Ba04646151_s1, Ba04646153_s1
Set Membership:
Recommended Assay
Microbe Detection
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline