Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ba00005009_po
Gene Symbol:
Bacteria
Gene Name:
Class A extended-spectrum beta-lactamase CTX-M
Gene Aliases:
Bacteria, blaCTX-M-180, blaCTX-M-181, blaCTX-M-182, blaCTX-M-183, blaCTX-M-184, blaCTX-M-185, blaCTX-M-186, blaCTX-M-187, blaCTX-M-188, blaCTX-M-189, blaCTX-M-19, blaCTX-M-190, blaCTX-M-191, blaCTX-M-192, blaCTX-M-193, blaCTX-M-194, blaCTX-M-195, blaCTX-M-196, blaCTX-M-199, blaCTX-M-2, blaCTX-M-20, blaCTX-M-200, blaCTX-M-201, blaCTX-M-202, blaCTX-M-203, blaCTX-M-204, blaCTX-M-205, blaCTX-M-206, blaCTX-M-207, blaCTX-M-208, blaCTX-M-209, blaCTX-M-21, blaCTX-M-210, blaCTX-M-211, blaCTX-M-212, blaCTX-M-213, blaCTX-M-214, blaCTX-M-215, blaCTX-M-216, blaCTX-M-217, blaCTX-M-218, blaCTX-M-219, blaCTX-M-22, blaCTX-M-220, blaCTX-M-221, blaCTX-M-222, blaCTX-M-223, blaCTX-M-224, blaCTX-M-225, blaCTX-M-226, blaCTX-M-227, blaCTX-M-228, blaCTX-M-229, blaCTX-M-23, blaCTX-M-230, blaCTX-M-231, blaCTX-M-232, blaCTX-M-233, blaCTX-M-235, blaCTX-M-236, blaCTX-M-237, blaCTX-M-238, blaCTX-M-239, blaCTX-M-24, blaCTX-M-240, blaCTX-M-241, blaCTX-M-242, blaCTX-M-243, blaCTX-M-244, blaCTX-M-245, blaCTX-M-25, blaCTX-M-26, blaCTX-M-27, blaCTX-M-28, blaCTX-M-29, blaCTX-M-3, blaCTX-M-30, blaCTX-M-31, blaCTX-M-32, blaCTX-M-33, blaCTX-M-34, blaCTX-M-35, blaCTX-M-36, blaCTX-M-37, blaCTX-M-38, blaCTX-M-39, blaCTX-M-4, blaCTX-M-40, blaCTX-M-41, blaCTX-M-42, blaCTX-M-43, blaCTX-M-44, blaCTX-M-46, blaCTX-M-47, blaCTX-M-48, blaCTX-M-49, blaCTX-M-5, blaCTX-M-50, blaCTX-M-51, blaCTX-M-52, blaCTX-M-53, blaCTX-M-54, blaCTX-M-55, blaCTX-M-56, blaCTX-M-58, blaCTX-M-59, blaCTX-M-6, blaCTX-M-60, blaCTX-M-61, blaCTX-M-62, blaCTX-M-63, blaCTX-M-64, blaCTX-M-65, blaCTX-M-66, blaCTX-M-67, blaCTX-M-68, blaCTX-M-69, blaCTX-M-7, blaCTX-M-71, blaCTX-M-72, blaCTX-M-73, blaCTX-M-76, blaCTX-M-77, blaCTX-M-78, blaCTX-M-79, blaCTX-M-8, blaCTX-M-80, blaCTX-M-82, blaCTX-M-83, blaCTX-M-84, blaCTX-M-85, blaCTX-M-86, blaCTX-M-87, blaCTX-M-88, blaCTX-M-89, blaCTX-M-9, blaCTX-M-90, blaCTX-M-91, blaCTX-M-92, blaCTX-M-93, blaCTX-M-94, blaCTX-M-95, blaCTX-M-96, blaCTX-M-97, blaCTX-M-98, blaCTX-M-99, blaCTX-M-1, blaCTX-M-10, blaCTX-M-100, blaCTX-M-101, blaCTX-M-102, blaCTX-M-103, blaCTX-M-104, blaCTX-M-105, blaCTX-M-110, blaCTX-M-111, blaCTX-M-112, blaCTX-M-113, blaCTX-M-114, blaCTX-M-115, blaCTX-M-116, blaCTX-M-117, blaCTX-M-12, blaCTX-M-121, blaCTX-M-122, blaCTX-M-123, blaCTX-M-124, blaCTX-M-125, blaCTX-M-126, blaCTX-M-127, blaCTX-M-129, blaCTX-M-13, blaCTX-M-130, blaCTX-M-131, blaCTX-M-134, blaCTX-M-136, blaCTX-M-137, blaCTX-M-138, blaCTX-M-139, blaCTX-M-14, blaCTX-M-140, blaCTX-M-141, blaCTX-M-142, blaCTX-M-143, blaCTX-M-144, blaCTX-M-146, blaCTX-M-147, blaCTX-M-148, blaCTX-M-15, blaCTX-M-150, blaCTX-M-152, blaCTX-M-153, blaCTX-M-154, blaCTX-M-155, blaCTX-M-156, blaCTX-M-157, blaCTX-M-158, blaCTX-M-159, blaCTX-M-16, blaCTX-M-160, blaCTX-M-161, blaCTX-M-162, blaCTX-M-163, blaCTX-M-164, blaCTX-M-165, blaCTX-M-166, blaCTX-M-167, blaCTX-M-168, blaCTX-M-169, blaCTX-M-17, blaCTX-M-170, blaCTX-M-171, blaCTX-M-172, blaCTX-M-173, blaCTX-M-174, blaCTX-M-176, blaCTX-M-177, blaCTX-M-178, blaCTX-M-179, class A extended-spectrum beta-lactamase (blaCTX-M), inhibitor-resistant class A extended-spectrum beta-lactamase (blaCTX-M), blaCTX-M
Chromosome Location:
-
Species:
Bacteria
Species Specific ID (Flybase ID):
-
Assay ID
Ba00005009_po
Best Coverage
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Updated Mapping Information
This pooled assay is composed of the following assays: Ba04646127_s1, Ba04646142_s1, Ba04646149_s1, Ba04646154_s1
Set Membership:
Recommended Assay
Microbe Detection
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline