Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ba00005033_po
Gene Symbol:
Bacteria
Gene Name:
Oxacillin-hydrolyzing class D beta-lactamase OXA; Class A extended-spectrum beta-lactamase GES
Gene Aliases:
Bacteria, blaGES-1, blaGES-10, blaGES-11, blaGES-12, blaGES-13, blaGES-14, blaGES-15, blaGES-16, blaGES-17, blaGES-18, blaGES-19, blaGES-2, blaGES-20, blaGES-21, blaGES-22, blaGES-24, blaGES-25, blaGES-26, blaGES-27, blaGES-28, blaGES-29, blaGES-3, blaGES-30, blaGES-31, blaGES-33, blaGES-34, blaGES-35, blaGES-36, blaGES-37, blaGES-38, blaGES-39, blaGES-4, blaGES-40, blaGES-42, blaGES-43, blaGES-45, blaGES-46, blaGES-47, blaGES-48, blaGES-5, blaGES-6, blaGES-7, blaGES-8, blaGES-9, blaOXA-1, blaOXA-224, blaOXA-31, blaOXA-320, blaOXA-392, blaOXA-4, blaOXA-47, blaOXA-534, blaOXA-675, blaOXA-796, blaOXA-921, OXA-1 family class D beta-lactamase (blaOXA), OXA-1 family class D extended-spectrum beta-lactamase (blaOXA), OXA-1 family oxacillin-hydrolyzing class D beta-lactamase (blaOXA), carbapenem-hydrolyzing class A beta-lactamase (blaGES), class A beta-lactamase (blaGES), class A extended-spectrum beta-lactamase (blaGES), inhibitor-resistant carbapenem-hydrolyzing class A beta-lactamase (blaGES), oxacillin-hydrolyzing class D beta-lactamase (blaOXA), blaGES,blaOXA
Chromosome Location:
-
Species:
Bacteria
Species Specific ID (Flybase ID):
-
Assay ID
Ba00005033_po
Best Coverage
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Updated Mapping Information
This pooled assay is composed of the following assays: Ba04646133_s1, Ba04646151_s1
Set Membership:
Recommended Assay
Microbe Detection
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-Offline